Learn More
CD70 Mouse anti-Human, PE, Clone: KI-24, BD
Mouse Monoclonal Antibody
$219.00 - $428.00
Specifications
Antigen | CD70 |
---|---|
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | PE |
Host Species | Mouse |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
BDB561935
|
BD Biosciences
561935 |
25 Tests |
Each for $219.00
|
|
|||||
BDB555835
|
BD Biosciences
555835 |
100 Tests |
Each for $428.00
|
|
|||||
Description
The Ki-24 monoclonal antibody specifically binds to human CD70. CD70 is a type II transmembrane glycoprotein and member of the TNF Superfamily. CD70 is also known as Tumor necrosis factor ligand superfamily member 7 (TNFSF7), CD27 ligand (CD27-L, CD27L, CD27LG), and KI-24 antigen. The CD70 antigen immunoprecipitates as five bands (50, 70, 90, 100 and 160 kDa) under non-reducing conditions. CD70 is strongly expressed on Reed-Sternberg cells, some activated T or B cells and Epstein Barr Virus (EBV)-positive lymphoblastoid cell lines. CD70 plays roles in the activation, proliferation and differentiation of B cells and T cells including the enhanced production of cytotoxic T cells.
Specifications
CD70 | |
Monoclonal | |
Mouse | |
Human | |
IgG3 κ | |
Affinity Purified |
Flow Cytometry | |
PE | |
RUO | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.