Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD71 Rat anti-Mouse, BUV496, Clone: C2 (also known as C2F2), BD Biosciences™

Rat Monoclonal Antibody

Supplier:  BD Biosciences 741066

Catalog No. BDB741066


Only null left
Add to Cart

Description

Description

CD71 Monoclonal antibody specifically detects CD71 in Mouse samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CD71
Monoclonal
Brilliant Ultraviolet 496
Transferrin Receptor; TR; TfR; TfR1; Tfrc; Trfr; Mtvr-1
Mouse cell line
RUO
Mouse
IgG1 κ
Flow Cytometry
C2 (also known as C2F2)
Aqueous buffered solution containing ≤0.09% sodium azide.
Rat
50 μg
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon BUV496 under optimal conditions that minimize unconjugated dye and antibody.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.