Learn More
Invitrogen™ CD71 (Transferrin Receptor) Monoclonal Antibody (OKT9 (OKT-9)), APC-eFluor™ 780, eBioscience™, Invitrogen™
Description
The OKT9 MAb reacts with human CD71, a 170-180 kDa type II transmembrane protein. CD71, the transferrin receptor, exists as a homodimer on the cell surface and is essential for cellular growth. CD71 is expressed by immature proliferating cells and at low levels on resting mature lymphocytes. Antigen or mitogen stimulation of T and B cells upregulates the expression of CD71.
Specifications
Specifications
| Antigen | CD71 (Transferrin Receptor) |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | OKT9 (OKT-9) |
| Concentration | 5 μL/Test |
| Conjugate | APC-eFluor 780 |
| Formulation | PBS with BSA and 0.09% sodium azide; pH 7.2 |
| Gene | TFRC |
| Gene Accession No. | P02786 |
| Gene Alias | 2610028K12Rik; AI195355; AI426448; AU015758; CD antigen CD71; CD71; chicken transferrin receptor; E430033M20Rik; I79_000642; IMD46; mammary tumor virus receptor 1; Mtvr1; Mtvr-1; p90; sTfR; T9; TfR; tfR1; TFR2; TFRC; TR; transferrin receptor; transferrin receptor (p90, CD71); transferrin receptor 2; transferrin receptor protein 1; Transferrin receptor protein 1, serum form; Trfr |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.