Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD71 (Transferrin Receptor) Rat anti-Mouse, BV750, Clone: C2 (also known as C2F2), BD Biosciences™
Rat Monoclonal Antibody
Supplier: BD Biosciences 747095
Description
CD71 (Transferrin Receptor) Monoclonal antibody specifically detects CD71 (Transferrin Receptor) in Mouse samples. It is validated for Flow Cytometry
Specifications
CD71 (Transferrin Receptor) | |
Monoclonal | |
Brilliant Violet 750 | |
Transferrin Receptor; TR; TfR; TfR1; Tfrc; Trfr; Mtvr-1 | |
Mouse cell line | |
RUO | |
Mouse | |
IgG1 κ |
Flow Cytometry | |
C2 (also known as C2F2) | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Rat | |
50 μg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon BV750 under optimal conditions that minimize unconjugated dye and antibody. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction