Learn More
CD72.1 Antibody (10-1.D.2), Biotin, Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NBP128024
Catalog No. NBP128024
Only null left
Add to Cart
Description
Description
CD72.1 Monoclonal specifically detects CD72.1 in Mouse samples. It is validated for Flow Cytometry, Immunoprecipitation, In vitro assay.
Specifications
Specifications
CD72.1
Monoclonal
0.5mg/mL
Flow Cytometry <= 1 ug/10^6 cells, Immunoprecipitation 1:10-1:500, In vitro assay
Cd72
DBA/2 mouse spleen cells
0.25 mg
Primary
Mouse CD72.1/Lyb-2.1, 45 kDa
Store at 4C. Do not freeze.
IgG2b κ
Flow Cytometry, Immunoprecipitation, In vitro Assay
10-1.D.2
Biotin
CD72 antigen, Ly19, Ly32, Lyb2, Lym19
Mouse
Protein A or G purified
RUO
12517
Mouse
Purified
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.
For Research Use Only