Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD72 Antibody (108), DyLight 755 , Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP306096IR
Description
CD72 Monoclonal antibody specifically detects CD72 in Mouse samples. It is validated for Flow CytometrySpecifications
CD72 | |
Monoclonal | |
DyLight 755 | |
CD72 antigenB-cell differentiation antigen CD72, CD72 molecule, CD72b, Lyb-2, LYB2lyb-2 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse CD72.1 (NP_001104530.1; Met1-Lys95). | |
0.1 mL | |
Adaptive Immunity, Immunology | |
971 | |
Store at 4°C in the dark. | |
IgG |
Flow Cytometry | |
108 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction