Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD72 Antibody (982405), DyLight 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB5405D
Description
CD72 Monoclonal antibody specifically detects CD72 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, CyTOFSpecifications
CD72 | |
Monoclonal | |
DyLight 350 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry, Immunocytochemistry, CyTOF | |
982405 | |
Flow Cytometry, Immunocytochemistry, CyTOF-ready | |
CD72 antigenB-cell differentiation antigen CD72, CD72 molecule, CD72b, Lyb-2, LYB2lyb-2 | |
Chinese hamster ovary cell line CHO-derived recombinant human CD72, Arg117-Asp359, Accession # P21854 | |
0.1 mL | |
Adaptive Immunity, Immunology | |
971 | |
Store at 4°C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction