Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
                                    CD72 Antibody, PE, Novus Biologicals™
 
                                    
                                    
                                    
                                    
                                
                            
                            
                            
                            
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10093559PE
Description
CD72 Polyclonal antibody specifically detects CD72 in Human samples. It is validated for Western BlotSpecifications
| CD72 | |
| Polyclonal | |
| Western Blot | |
| CD72 antigenB-cell differentiation antigen CD72, CD72 molecule, CD72b, Lyb-2, LYB2lyb-2 | |
| Synthetic peptide corresponding to residues 25-37 [LGQDPGADDDGEI] of the 42 kDa human CD72 protein. | |
| 0.1 mL | |
| Adaptive Immunity, Immunology | |
| 971 | |
| Store at 4C in the dark. | |
| IgG | 
| Western Blot | |
| PE | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified | 
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction
            