Learn More
CD72 Mouse anti-Human, BUV395, Clone: J4-117, BD Optibuild™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 743800
Description
The J4-117 monoclonal antibody specifically recognizes CD72. CD72 is a 39-43 kDa type II integral membrane protein that belongs to the C-type lectin superfamily. CD72 is expressed on B cells from early precursor stages through mature B cells. CD72 is not expressed on plasma cells. CD72 is also expressed on follicular dendritic reticulum cells and some macrophages. CD72 has been reported to bind to CD5, a glycoprotein expressed on all mature T cells and a subpopulation of B cells, suggesting a role in regulating T- and B-cell activation and proliferation.
The antibody was conjugated to BD Horizon™ BUV395 which is part of the BD Horizon Brilliant™ Ultraviolet family of dyes. This dye has been exclusively developed by BD Biosciences to have minimal spillover into other detectors, making it an optimal choice for multicolor flow cytometry. With an Ex Max at 348 nm and an Em Max at 395 nm, BD Horizon BUV395 can be excited with a 355 nm laser and detected with a 379/28 filter. BUV395 is a UV-excitable dye that has been developed exclusively by BD Biosciences. With an excitation max of 348 nm and emission max of 395 nm, BUV395 can be excited by the 355nm laser and detected in a 379/28 filter. This dye is optimal for multicolor flow cytometry because it has little to no spillover into other detectors.
Specifications
CD72 | |
J4-117 | |
Brilliant Ultraviolet 395 | |
Lyb-2, LYB2 | |
Affinity Purified | |
RUO | |
Human | |
IgG2b κ |
Monoclonal | |
0.2mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse | |
50 μg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.