Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD72 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597292
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597292 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597292 Supplier Invitrogen™ Supplier No. PA597292
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QVSQQLQQTN RVLEVTNSSL RQQLRLKITQ LGQSAEDLQG SRRELAQSQE ALQVEQRAHQ AAEGQLQACQ ADRQKTKETL QSEEQQRRAL EQKLSNMENR LKPFFTCGSA DTCCPSGWIM HQKSCFYISL TSKNWQESQK QCETLSSKLA TFSEIYPQSH SYYFLNSLLP NGGSGNSYWT GLSSNKDWKL TDDTQRTRTY AQSSKCNKVH KTWSWWTLES ESCRSSLPYI CEMTAFRFPD; Positive Samples: B-cell, Mouse liver, Mouse kidney, Rat liver, Rat kidney; Cellular Location: Membrane, Single-pass type II membrane protein.

CD72 is a transmembrane glycoprotein expressed as a homodimer especially in B cells, but also in other antigen presenting cells such as dendritic cells and macrophages. Through one of its immunoreceptor tyrosine-based inhibitory motives (ITIMs), CD72 interacts with tyrosine phosphatase SHP-1, thereby suppressing B cell responsiveness. Binding of CD72 with its ligand CD100 (Sema4D) prevents BCR association and phosphorylation of CD72 and results in dissociation of SHP-1 from CD72, thus enables B cell activation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD72
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.28 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene CD72
Gene Accession No. P21854, P21855
Gene Alias B-cell differentiation antigen CD72; CD72; CD72 antigen; Cd72 molecule; CD72b; CD72c; Ly-19; Ly32; Ly-32; LYB2; Lyb-2; Ly-m19; lymphocyte antigen 32; RP11-331F9.9
Gene Symbols CD72
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 120-359 of human CD72 (NP_0017731).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12517, 313498, 971
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.