Learn More
CD73 Mouse anti-Human, Brilliant Ultraviolet 737, Clone: AD2, BD Horizon™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 612813
Description
The AD2 monoclonal antibody specifically binds to ecto-5’-nucleotidase, a 70 kDa, glycosyl phosphatidylinositol (GPI)-anchored glycoprotein. CD73 is expressed on subsets of T and B lymphocytes, follicular dendritic cells, epithelial cells, endothelial cells and mesenchymal stem cells. Its expression on lymphocytes increases during T and B cell development. CD73 has enzymatic activity and catalyzes the dephosphorylation of adenosine monophosphate (AMP) converting it to adenosine. It has been suggested that CD73 can mediate costimulatory signals in T cell activation and adhesion of lymphocytes to endothelium.
The antibody was conjugated to BD Horizon BUV737 which is part of the BD Horizon Brilliant™ Ultraviolet family of dyes. This dye is a tandem fluorochrome of BD Horizon BUV395 with an Ex Max of 348-nm and an acceptor dye with an Em Max at 737-nm. BD Horizon Brilliant BUV737 can be excited by the ultraviolet laser (355 nm) and detected with a 740/35 filter. Due to the excitation of the acceptor dye by other laser lines, there may be significant spillover into channels detecting Alexa Fluor™ 700-like dyes (eg, 712/20-nm filter).

Specifications
CD73 | |
Monoclonal | |
Brilliant Ultraviolet 737 | |
Mouse | |
Affinity Purified | |
RUO | |
Human | |
IgG1 κ |
Flow Cytometry | |
AD2 | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Pre-B leukemia cell line | |
25 Tests | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.