Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD74 Antibody (6D9), DyLight 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP259698UV
Description
CD74 Monoclonal specifically detects CD74 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
CD74 | |
Monoclonal | |
DyLight 350 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
CD74 | |
Synthetic peptide corresponding to the N-terminal (1-100 aa) of human CD74 conjugated to KLH. Overlaps 100% with all 3 isoforms | |
100 μL | |
Primary | |
Detects approx 50kDa. No cross-reactivity against rat Cab Beta1b, Cav beta2a, and Cav betas3 in transfected cells. | |
Store at 4C in the dark. | |
IgG1 |
Western Blot, Immunocytochemistry, Immunofluorescence | |
6D9 | |
Western Blot, Immunocytochemistry/Immunofluorescence | |
CD74 antigen, CD74 antigen (invariant polypeptide of major histocompatibility complex, classII antigen-associated), CD74 molecule, major histocompatibility complex, class II invariant chain, DHLAGgamma chain of class II antigens, HLA class II histocompatibility antigen gamma chain, HLADG, HLA-DR antigens-associated invariant chain, HLA-DR-gamma, Ia antigen-associated invariant chain, Ia-associated invariant chain, Ia-GAMMA, Ii, MHC HLA-DR gamma chain, p33 | |
Mouse | |
Protein G purified | |
Immunology | |
972 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction