Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD74 Antibody (829706) [FITC], Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals FAB7478F
Description
CD74 Monoclonal antibody specifically detects CD74 in Mouse samples. It is validated for Flow Cytometry, CyTOFSpecifications
CD74 | |
Monoclonal | |
FITC | |
PBS | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Mouse | |
Purified |
Flow Cytometry, CyTOF | |
829706 | |
Flow Cytometry, CyTOF-ready | |
CD74 antigen, CD74 antigen (invariant polypeptide of major histocompatibility complex, classII antigen-associated), CD74 molecule, major histocompatibility complex, class II invariant chain, DHLAGgamma chain of class II antigens, HLA class II histocompatibility antigen gamma chain, HLADG, HLA-DR antigens-associated invariant chain, HLA-DR-gamma, Ia antigen-associated invariant chain, Ia-associated invariant chain, Ia-GAMMA, Ii, MHC HLA-DR gamma chain, p33 | |
Chinese hamster ovary cell line CHO-derived recombinant mouse CD74, Gln56-Cys215, Accession # P04441 | |
0.1 mL | |
Immunology | |
972 | |
Store at 4°C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction