Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD74 Antibody (CLIP/1133), Novus Biologicals™

Mouse Monoclonal Antibody
$235.64 - $529.81
Specifications
| Antigen | CD74 |
|---|---|
| Clone | CLIP/1133 |
| Concentration | 0.2 mg/mL |
| Dilution | Western Blot 0.5 - 1.0 ug/ml, Flow Cytometry 0.5 - 1 ug/million cells in 0.1 ml, Immunofluorescence 1 - 2 ug/ml |
| Applications | Western Blot, Flow Cytometry, Immunofluorescence |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP24499100
![]() |
Novus Biologicals
NBP2449910.02MG |
0.02 mg |
Each for $235.64
|
|
|||||
NBP24499101
![]() |
Novus Biologicals
NBP2449910.1MG |
0.1 mg |
Each for $529.81
|
|
|||||
NBP24499102
![]() |
Novus Biologicals
NBP2449910.2MG |
0.2 mg | N/A | N/A | N/A | ||||
Description
Ensure accurate, reproducible results in Western Blot, Flow Cytometry, Immunofluorescence
CD74 Monoclonal specifically detects CD74 in Human samples. It is validated for Western Blot.Specifications
| CD74 | |
| 0.2 mg/mL | |
| Western Blot, Flow Cytometry, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Immunology | |
| 10mM PBS and 0.05% BSA with 0.05% Sodium Azide | |
| 972 | |
| Recombinant full-length human CD74 protein | |
| Primary | |
| Store at 4C. |
| CLIP/1133 | |
| Western Blot 0.5 - 1.0 ug/ml, Flow Cytometry 0.5 - 1 ug/million cells in 0.1 ml, Immunofluorescence 1 - 2 ug/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CD74 antigen, CD74 antigen (invariant polypeptide of major histocompatibility complex, classII antigen-associated), CD74 molecule, major histocompatibility complex, class II invariant chain, DHLAGgamma chain of class II antigens, HLA class II histocompatibility antigen gamma chain, HLADG, HLA-DR antigens-associated invariant chain, HLA-DR-gamma, Ia antigen-associated invariant chain, Ia-associated invariant chain, Ia-GAMMA, Ii, MHC HLA-DR gamma chain, p33 | |
| CD74 | |
| IgG1 κ | |
| Protein A or G purified | |
| Recognizes proteins of 33, 35 and 41kDa, which are identified as various isoforms of CD74. Its epitope is localized in the extracellular domain of CD74. CD74 is a type II transmembrane protein which binds to the peptide binding groove of newly synthesized MHC class II alpha/beta heterodimers and prevents their premature association with endogenous polypeptides. The CD74 molecule plays a critical role in the presentation of peptides, by the MHC class II antigens, to CD4 positive lymphocytes. CD74 is expressed on MHC class II positive cells including B cells, a subset of activated T cells, monocytes, and dendritic cells and by various types of carcinomas. CD74 is expressed primarily by antigen presenting cells, such as B-lymphocytes (from before the pre-B cell stage to before the plasma cell stage), macrophages, and monocytes, and many epithelial cells. Anti-CD74 stains predominantly germinal center lymphocytes and B-cell lymphomas, but rarely T-cell lymphomas. Anti-CD74 has been shown to be useful in differentiating atypical fibroxanthoma (-) from malignant fibrous histiocytoma (+). |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title