Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD74 Antibody (PIN.1), PE/Cy5.5, Novus Biologicals™
SDP

Catalog No. N101985PEC5 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N101985PEC5 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N101985PEC5 Supplier Novus Biologicals Supplier No. NB1001985PECY55
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD74 Monoclonal specifically detects CD74 in Human, Mouse (Negative) samples. It is validated for Flow Cytometry.

Specifications

Antigen CD74
Applications Flow Cytometry
Classification Monoclonal
Clone PIN.1
Conjugate PE-Cyanine5.5
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias CD74 antigen, CD74 antigen (invariant polypeptide of major histocompatibility complex, classII antigen-associated), CD74 molecule, major histocompatibility complex, class II invariant chain, DHLAGgamma chain of class II antigens, HLA class II histocompatibility antigen gamma chain, HLADG, HLA-DR antigens-associated invariant chain, HLA-DR-gamma, Ia antigen-associated invariant chain, Ia-associated invariant chain, Ia-GAMMA, Ii, MHC HLA-DR gamma chain, p33
Gene Symbols CD74
Host Species Mouse
Immunogen Peptide corresponding to residues 12-27 of CD74, human invariant chain (short form).
Purification Method Protein G purified
Quantity 0.1 mL
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 972.0
Test Specificity This detects an ∽33-35 kDa protein doublet, corresponding to the apparent molecular mass of the p33 and p35 forms of human CD74.
Target Species Human, Mouse (Negative)
Content And Storage Store at 4°C in the dark. Do not freeze.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.