Learn More
Invitrogen™ CD74 Monoclonal Antibody (LN2), Biotin

Description
CD74 Monoclonal Antibody for Flow
Specifications
Specifications
| Antigen | CD74 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | LN2 |
| Concentration | 0.1 mg/mL |
| Conjugate | Biotin |
| Formulation | PBS with <0.1% sodium azide; pH 7.4 |
| Gene | CD74 |
| Gene Accession No. | P04233 |
| Gene Alias | CD74; CD74 antigen; CD74 antigen (invariant polypeptide of major histocompatibility complex, class II antigen-associated); CD74 molecule; Cd74 molecule, major histocompatibility complex, class II invariant chain; class II-associated invariant chain peptide; CLIP; DHLAG; dinucleotide microsatellite; gamma chain of class II antigens; H-2 class II histocompatibility antigen gamma chain; histocompatibility: class II antigens, gamma chain of; HLA class II histocompatibility antigen gamma chain; HLADG; HLA-DR antigens-associated invariant chain; HLA-DR-gamma; ia antigen-associated invariant chain; Ia-associated invariant chain; Ia-GAMMA; Ii; invariant polypeptide of major histocompatibility complex, class II antigen-associated; INVG34; MHC class II-associated invariant chain; MHC HLA-DR gamma chain; p33 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.