Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD74 Monoclonal Antibody (OTI1G10), TrueMAB™, OriGene
SDP

Catalog No. 501677068
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-167-7068 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-167-7068 Supplier OriGene Supplier No. TA507336
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Expression of the invariant chain of the HLA-DR antigen (CD74) by B-lymphocytes begins in the pre-B stage but lost before the final differentiation to plasma cells. This protein is expressed in B-lymphocytes and many B-cell lymphomas.

Specifications

Antigen CD74
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI1G10
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene CD74
Gene Accession No. P04233
Gene Alias DHLAG, HLADG, II, Ia-GAMMA
Gene Symbols CD74
Host Species Mouse
Immunogen Full length human recombit protein of human CD74 produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 972
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.