Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD74 Monoclonal Antibody (PIN.1), FITC
GREENER_CHOICE

Catalog No. PIMA545125
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA545125 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA545125 Supplier Invitrogen™ Supplier No. MA545125
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

1 μg/mL of MA5-45125 was sufficient for detection of CD74 in 20 μg of PALA cell lysates by colorimetric immunolot analysis using goat anti-mouse IgG: AP as the secondary antibody. Detects approximately 33-35kDa protein doublet corresponding to the molecular mass of the p33 and p35 forms of human CD74.

HLA-DR, like other MHC class II molecules, is a transmembrane glycoprotein composed of a 36 kDa alpha chain (DRA) and 27 kDa beta chain (DRB). The alpha chain gene contains 5 exons. Exon 1 encodes the leader peptide, exons 2 and 3 encode the two extracellular domains, and exon 4 encodes the transmembrane domain and the cytoplasmic tail. DRA does not have polymorphisms in the peptide binding part and acts as the sole alpha chain for DRB1, DRB3, DRB4 and DRB5. Within the DR molecule the beta chain contains all the polymorphisms specifying the peptide binding specificities. Hundreds of DRB1 alleles have been described and typing for these polymorphisms is routinely done for bone marrow and kidney transplantation. HLA-DR is expressed primarily on antigen presenting cells such as B lymphocytes, monocytes, macrophages, thymic epithelial cells and activated T lymphocytes. Three loci, DR, DQ and DP, encode the major expressed products of the human class II region. The human MHC class II molecules bind intracellularly processed peptides, present them to T-helper cells, and have a critical role in the initiation of the immune response.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD74
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone PIN.1
Concentration 1 mg/mL
Conjugate FITC
Formulation PBS, 9.09mM sodium bicarbonate with 640.91mM DMSO, 136.36mM ethanolamine and no preservative; pH 7.2
Gene CD74
Gene Accession No. P04233, P04441
Gene Alias CD74; CD74 antigen; CD74 antigen (invariant polypeptide of major histocompatibility complex, class II antigen-associated); CD74 molecule; Cd74 molecule, major histocompatibility complex, class II invariant chain; class II-associated invariant chain peptide; CLIP; DHLAG; dinucleotide microsatellite; gamma chain of class II antigens; H-2 class II histocompatibility antigen gamma chain; histocompatibility: class II antigens, gamma chain of; HLA class II histocompatibility antigen gamma chain; HLADG; HLA-DR antigens-associated invariant chain; HLA-DR-gamma; ia antigen-associated invariant chain; Ia-associated invariant chain; Ia-GAMMA; Ii; invariant polypeptide of major histocompatibility complex, class II antigen-associated; INVG34; MHC class II-associated invariant chain; MHC HLA-DR gamma chain; p33
Gene Symbols CD74
Host Species Mouse
Immunogen Human CD74 invariant chain synthetic peptide.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16149, 972
Target Species Human, Mouse
Content And Storage 4°C, store in dark
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.