Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD74 Rabbit anti-Human, Mouse, Rat, Clone: LN-2, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP3120790.2MG
Description
CD74 Monoclonal antibody specifically detects CD74 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence, ImmunoprecipitationSpecifications
CD74 | |
Monoclonal | |
1 mg/mL | |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation | |
CD74 antigen, CD74 antigen (invariant polypeptide of major histocompatibility complex, classII antigen-associated), CD74 molecule, major histocompatibility complex, class II invariant chain, DHLAGgamma chain of class II antigens, HLA class II histocompatibility antigen gamma chain, HLADG, HLA-DR antigens-associated invariant chain, HLA-DR-gamma, Ia antigen-associated invariant chain, Ia-associated invariant chain, Ia-GAMMA, Ii, MHC HLA-DR gamma chain, p33 | |
This antibody was raised by immunizing mice with SU-DHL-4 lymphoma cells. | |
0.2 mg | |
Immunology | |
972 | |
Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C. |
Immunohistochemistry | |
LN-2 | |
Unconjugated | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Rat | |
IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction