Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD74 Rabbit anti-Human, Unconjugated, Clone: CLIP/3127R, Abnova™

Catalog No. 89339717 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-339-717 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-339-717 Supplier Abnova Corporation Supplier No. RAB00678
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody.

CD74 Monoclonal antibody specifically detects CD74 in Baboon, Human, Mouse samples. It is validated for IHC-P, Flow Cyt.

Specifications

Antigen CD74
Applications Immunohistochemistry (Paraffin), Flow Cytometry
Classification Monoclonal
Clone CLIP/3127R
Conjugate Unconjugated
Description CD74 molecule, major histocompatibility complex, class II invariant chain
Gene CD74
Gene Alias DHLAG, HLADG, Ia-GAMMA
Host Species Rabbit
Immunogen SU-DHL-4 lymphoma cells
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 972
Target Species Baboon, Human, Mouse
Content And Storage Store at -20 to -80°C. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.