Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD79A Antibody (IGA/1406), Novus Biologicals™
SDP

Mouse Monoclonal Antibody

$268.00 - $524.00

Specifications

Antigen CD79A
Clone IGA/1406
Concentration 0.2 mg/mL
Dilution Western Blot 0.5 - 1 ug/ml, Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 0.5 - 1 ug/ml
Applications Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB25325420
SDP
View Documents
Novus Biologicals
NBP25325420UG
20 μg
Each for $268.00
Only null left
Add to Cart
 
NB253254100
SDP
View Documents
Novus Biologicals
NBP253254100UG
100 μg
Each for $524.00
Only null left
Add to Cart
 
NB253254200
SDP
View Documents
Novus Biologicals
NBP253254200UG
200 μg N/A N/A N/A
Description

Description

CD79A Monoclonal specifically detects CD79A in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

CD79A
0.2 mg/mL
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence
Unconjugated
Mouse
Adaptive Immunity, Cell Biology, Immunology, Phospho Specific
10mM PBS with 0.05% BSA with 0.05% Sodium Azide
973
A synthetic peptide corresponding to aa 202-216 (GTYQDVGSLNIADVQ) of human CD79a protein.
Primary
Store at 4C.
IGA/1406
Western Blot 0.5 - 1 ug/ml, Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 0.5 - 1 ug/ml
Monoclonal
Purified
RUO
Human
CD79a antigen, CD79A antigen (immunoglobulin-associated alpha), CD79a molecule, immunoglobulin-associated alpha, IGAB-cell antigen receptor complex-associated protein alpha chain, Ig-alpha, MB1, MB-1, MB-1 membrane glycoprotein, Membrane-bound immunoglobulin-associated protein, Surface IgM-associated protein
CD79A
IgG3
Protein A or G purified
35 kDa
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.