Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD79A Antibody (IGA/1790R), Allophycocyanin , Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP311617APC
Description
CD79A Monoclonal antibody specifically detects CD79A in Human, Mouse, Rat, Porcine, Bovine, Canine, Monkey samples. It is validated for Flow CytometrySpecifications
CD79A | |
Monoclonal | |
APC | |
CD79a antigen, CD79A antigen (immunoglobulin-associated alpha), CD79a molecule, immunoglobulin-associated alpha, IGAB-cell antigen receptor complex-associated protein alpha chain, Ig-alpha, MB1, MB-1, MB-1 membrane glycoprotein, Membrane-bound immunoglobulin-associated protein, Surface IgM-associated protein | |
A synthetic peptide corresponding to aa 202-216 (GTYQDVGSLNIADVQ) of human CD79A protein. (Uniprot: P11912) | |
0.1 mL | |
Adaptive Immunity, Cell Biology, Immunology, Phospho Specific | |
973 | |
Store at 4°C in the dark. | |
IgG |
Flow Cytometry | |
IGA/1790R | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human, Mouse, Rat, Pig, Bovine, Canine, Monkey | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction