Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD79A Antibody (JCB117 + HM47/A9), PE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP234653PE
Description
CD79A Monoclonal specifically detects CD79A in Human, Mouse, Rat, Porcine, Bovine, Monkey samples. It is validated for Flow Cytometry.Specifications
| CD79A | |
| Monoclonal | |
| PE | |
| CD79a antigen, CD79A antigen (immunoglobulin-associated alpha), CD79a molecule, immunoglobulin-associated alpha, IGAB-cell antigen receptor complex-associated protein alpha chain, Ig-alpha, MB1, MB-1, MB-1 membrane glycoprotein, Membrane-bound immunoglobulin-associated protein, Surface IgM-associated protein | |
| Mouse | |
| 44 kDa | |
| 0.1 mL | |
| Adaptive Immunity, Cell Biology, Immunology | |
| 973 | |
| Human, Mouse, Rat, Pig, Bovine, Primate | |
| Purified |
| Flow Cytometry | |
| JCB117 + HM47/A9 | |
| Flow Cytometry | |
| CD79A | |
| A synthetic peptide corresponding to aa 202-216 (GTYQDVGSLNIADVQ) of human CD79A protein (JCB117 and HM47/A9). (Uniprot: P11912) | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| A disulphide-linked heterodimer, consisting of mb-1 (or CD79a) and B29 (or CD79b) polypeptides, is non-covalently associated with membrane-bound immunoglobulins on B cells. This complex of mb-1 and B29 polypeptides and immunoglobulin constitute the B cell Ag receptor. CD79a first appears at pre B cell stage, early in maturation, and persists until the plasma cell stage where it is found as an intracellular component. CD79a is found in the majority of acute leukemias of precursor B cell type, in B cell lines, B cell lymphomas, and in some myelomas. It is not present in myeloid or T cell lines. Anti-CD79a is generally used to complement anti-CD20 especially for mature B-cell lymphomas after treatment with Rituximab (anti-CD20). This antibody will stain many of the same lymphomas as anti-CD20, but also is more likely to stain B-lymphoblastic lymphoma/leukemia than is anti-CD20. Anti-CD79a also stains more cases of plasma cell myeloma and occasionally some types of endothelial cells as well. | |
| Store at 4C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction