Learn More
Epredia™ Lab Vision™ CD79a/mb-1 (B-Cell Marker), Rabbit Monoclonal Antibody
Shop All Epredia ProductsDescription
Host Species: Rabbit
Clone: SP18
Species Reactivity: Human. Does not react with mouse, rat, canine, monkey and opossum
Epitope: N-terminus
Immunogen: Synthetic peptide derived from N-terminal region of human CD79a protein
Molecular Weight: 50 kDa
Positive Control: Tonsil
Cellular Localization: Membrane
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
Specifications
| Antigen | CD79a/mb-1 (B-Cell Marker) |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | SP18 |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Immunogen | Synthetic peptide derived from N-terminal region of human CD79a protein |
| Quantity | 12 mL |
| Regulatory Status | IVD |
| Research Discipline | Cancer and Tumor Biology |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.