Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ CD79a/mb-1 (B-Cell Marker), Rabbit Monoclonal Antibody

Catalog No. RM9118S1 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
500 μL
1 mL
7 mL
12 mL
Form:
Concentrated
Prediluted
Ready-to-Use
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form
RM9118S1 500 μL Concentrated
RM9118S0 100 μL Concentrated
RM9118S 1 mL Concentrated
RM9118R1 1 mL Prediluted
RM9118R7 7 mL Prediluted
RM9118RQ 12 mL Ready-to-Use
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. RM9118S1 Supplier Epredia™ Supplier No. RM9118S1

Obtain accurate, reproducible results in immunohistochemistry experiments with Epredia™ CD79a/mb-1 (B-Cell Marker), Rabbit Monoclonal Antibody.

A disulphide-linked heterodimer, consisting of CD79a/mb-1 and CD79b/B29 polypeptides, is non-covalently associated with membrane-bound immunoglobulins on B cells to constitute the B cell Ag receptor. CD79a first appears at pre B cell stage and persists until the plasma cell stage where it is found as an intracellular component. CD79a is found in the majority of acute leukemias of precursor B cell type, in B cell lines, B cell lymphomas, and in some myelomas.

Host Species: Rabbit

Clone: SP18

Species Reactivity: Human. Does not react with mouse, rat, canine, monkey and opossum

Epitope: N-terminus

Immunogen: Synthetic peptide derived from N-terminal region of human CD79a protein

Molecular Weight: 50 kDa

Positive Control: Tonsil

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Specifications

Antigen CD79a/mb-1 (B-Cell Marker)
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone SP18
Conjugate Unconjugated
Host Species Rabbit
Immunogen Synthetic peptide derived from N-terminal region of human CD79a protein
Quantity 500 μL
Regulatory Status IVD
Research Discipline Cancer and Tumor Biology
Primary or Secondary Primary
Target Species Human
Form Concentrated
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.