Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD79a Monoclonal Antibody (24C2.5), FITC, eBioscience™

Mouse Monoclonal Antibody

Supplier:  Invitrogen™ 11079180

Encompass_Preferred

Catalog No. 50-112-2079


Only null left
Add to Cart

Description

Description

Description: This 24C2.5 monoclonal antibody reacts with the intracellular tail of mouse CD79a (also known as Ig alpha or mb-1), a signaling component of the B cell receptor (BCR). CD79a heterodimerizes with CD79b (Ig beta, B29), and together with surface Ig, make up the BCR. Both CD79 subunits consist of a single extracellular Ig domain, a transmembrane domain, and an intracellular signaling domain. CD79a is expressed almost exclusively on B cells, as well as in most B lineage acute lymphoblastic leukemias. The cytoplasmic domain of CD79a contains immunoreceptor tyrosine-based activation motifs (ITAMs), which constitute the signal transducing portions of the BCR. Tyrosine phosphorylation of these ITAMs by Syk and Lyn initiates numerous signaling cascades, resulting in B cell activation, proliferation, and differentiation. Applications Reported: This 24C2.5 antibody has been reported for use in intracellular staining followed by flow cytometric analysis. Applications Tested: This 24C2.5 antibody has been tested by flow cytometric analysis on mouse splenocytes. This can be used at less than or equal to 1 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 488 nm; Emission: 520 nm; Laser: Blue...

CD79a is a disulphide-linked heterodimer that includes B29 (CD79b) polypeptide. CD79a is a B lymphocyte antigen receptor with an antigen-specific surface component Ig (immunoglobulin) that associates with Ig-alpha and Ig-beta, necessary elements for the expression and function of the B-cell antigen receptor. CD79a first appears at pre-B cell stage and persists until the plasma cell stage where it is found as an intracellular component. CD79a is found in the majority of acute leukemias of precursor B cell type, in B cell lines, B cell lymphomas, and in some myelomas. It is not present in myeloid or T cell lines. Diseases associated with CD79a include Agammaglobulinemia 3 and Agammaglobulinemia, Non-Bruton type.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CD79a
Monoclonal
0.5 mg/mL
PBS with 0.09% sodium azide; pH 7.2
P11911
CD79A
Affinity Chromatography
RUO
12518
4°C, store in dark, DO NOT FREEZE!
Liquid
Flow Cytometry
24C2.5
FITC
CD79A
B-cell antigen receptor; B-cell antigen receptor complex-associated protein alpha chain; B-cell antigen receptor complex-associated protein alpha chain-like; CD79; CD79a; CD79A antigen (immunoglobulin-associated alpha); CD79a molecule; CD79a molecule, immunoglobulin-associated alpha; CD79A protein; CD79-alpha protein; EGK_10665; Ig alpha; IGA; Igalpha; ig-alpha; immunoglobulin-associated alpha; LOC100913063; Ly54; Ly-54; MB1; MB-1; MB-1 membrane glycoprotein; Membrane-bound immunoglobulin-associated protein; surface IgM-associated protein
Mouse
25 μg
Primary
Mouse
Antibody
IgG1 κ
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Reproductive Harm - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.