Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD79a Monoclonal Antibody (HM47/A9), Biotin, Invitrogen™

Catalog No. PIMA513209
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA513209 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA513209 Supplier Thermo Scientific Supplier No. MA513209

Mouse Monoclonal Antibody

MA5-13209 targets CD79a in IF, IHC (P) and WB applications and shows reactivity with Bovine, Equine, Guinea Pig, Human, mouse, Non-human primate, Porcine, Rabbit, chicken and Rat samples. The MA5-13209 immunogen is a synthetic peptide consisting of GTYQDVGSLNIADVQ of human CD79a protein.

CD79a Monoclonal antibody specifically detects CD79a in Bovine, Chicken, Equine, Guinea Pig, Human, Mouse, Non-human Primate, Porcine, Rabbit, Rat samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD79a
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone HM47/A9
Concentration 0.2 mg/mL
Conjugate Biotin
Formulation PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4
Gene CD79A
Gene Accession No. P11911, P11912, P40293
Gene Alias B-cell antigen receptor; B-cell antigen receptor complex-associated protein alpha chain; B-cell antigen receptor complex-associated protein alpha chain-like; CD79; CD79a; CD79A antigen (immunoglobulin-associated alpha); CD79a molecule; CD79a molecule, immunoglobulin-associated alpha; CD79A protein; CD79-alpha protein; EGK_10665; Ig alpha; IGA; Igalpha; ig-alpha; immunoglobulin-associated alpha; LOC100913063; Ly54; Ly-54; MB1; MB-1; MB-1 membrane glycoprotein; Membrane-bound immunoglobulin-associated protein; surface IgM-associated protein
Gene Symbols CD79A
Host Species Mouse
Immunogen A synthetic peptide consisting of GTYQDVGSLNIADVQ of human CD79a protein
Purification Method Protein G
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100052219, 100190992, 100724815, 100913063, 12518, 281674, 973
Target Species Bovine, Chicken, Equine, Guinea Pig, Human, Mouse, Non-human Primate, Pig, Rabbit, Rat
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.