Learn More
Invitrogen™ CD79a Monoclonal Antibody (ZL7-4), FITC

Description
For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL. For FACS testing, we recommend incubation times of at least 30 minutes with this antibody. Mouse anti Human CD79a antibody, clone ZL7-4 recognizes the human B-cell antigen receptor complex-associated protein alpha chain, also known as MB-1 membrane glycoprotein or CD79a.
Specifications
Specifications
| Antigen | CD79a |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | ZL7-4 |
| Concentration | 0.1 mg/mL |
| Conjugate | FITC |
| Formulation | PBS with 1% BSA and 0.09% sodium azide; pH 7.4 |
| Gene | CD79A |
| Gene Accession No. | P11912 |
| Gene Alias | B-cell antigen receptor; B-cell antigen receptor complex-associated protein alpha chain; B-cell antigen receptor complex-associated protein alpha chain-like; CD79; CD79a; CD79A antigen (immunoglobulin-associated alpha); CD79a molecule; CD79a molecule, immunoglobulin-associated alpha; CD79A protein; CD79-alpha protein; EGK_10665; Ig alpha; IGA; Igalpha; ig-alpha; immunoglobulin-associated alpha; LOC100913063; Ly54; Ly-54; MB1; MB-1; MB-1 membrane glycoprotein; Membrane-bound immunoglobulin-associated protein; surface IgM-associated protein |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.