Learn More
Invitrogen™ CD79a Monoclonal Antibody (HM47), PerCP-Cyanine5.5, eBioscience™
Mouse Monoclonal Antibody
Supplier: Invitrogen™ 45079242

Description
Description: The monoclonal antibody HM47 recognizes the cytoplasmic domain of CD79a, also known as mb-1. CD79a is a 47 kDa membrane glycoprotein that associates with CD79b to form the heterodimeric B cell receptor (BCR). This receptor is responsible for B cell signaling, resulting in activation, apoptosis or anergy. Expression of CD79a is found throughout development from the earliest pre-B cell to plasma cells. CD79 is expressed on B cell neoplasms and some non-B cell malignancies such as AML. Unlike other antibodies that can be masked in B cell neoplastic cells, HM47 is a reliable antibody for recognizing CD79a and, as such, is a good marker for B cells. Because several variants of CD79a exist, the exact location of the epitope may differ slightly. Nevertheless, in humans thie epitope is typically amino acids 208-222. The HM47 antibody reacts to human, mouse, rat, dog, primates (rhesus, chimpanzee, macaque and baboon) pig, guinea rabbit, pig, horse, cow and chicken. Applications Reported: This HM47 antibody has been reported for use in intracellular staining followed by flow cytometric analysis. Applications Tested: This HM47 antibody has been pre-titrated and tested by intracellular staining and flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.03 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but...
CD79a is a disulphide-linked heterodimer that includes B29 (CD79b) polypeptide. CD79a is a B lymphocyte antigen receptor with an antigen-specific surface component Ig (immunoglobulin) that associates with Ig-alpha and Ig-beta, necessary elements for the expression and function of the B-cell antigen receptor. CD79a first appears at pre-B cell stage and persists until the plasma cell stage where it is found as an intracellular component. CD79a is found in the majority of acute leukemias of precursor B cell type, in B cell lines, B cell lymphomas, and in some myelomas. It is not present in myeloid or T cell lines. Diseases associated with CD79a include Agammaglobulinemia 3 and Agammaglobulinemia, Non-Bruton type.
Specifications
CD79a | |
Monoclonal | |
5 μL/Test | |
PBS with BSA and 0.09% sodium azide; pH 7.2 | |
P0CAN6, P11911, P11912, P40293 | |
CD79A | |
Affinity chromatography | |
RUO | |
100052219, 100190992, 100724815, 100913063, 12518, 281674, 484483, 722190, 740560, 973 | |
4°C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
HM47 | |
PerCP-Cyanine5.5 | |
CD79A | |
B-cell antigen receptor; B-cell antigen receptor complex-associated protein alpha chain; B-cell antigen receptor complex-associated protein alpha chain-like; CD79; CD79a; CD79A antigen (immunoglobulin-associated alpha); CD79a molecule; CD79a molecule, immunoglobulin-associated alpha; CD79A protein; CD79-alpha protein; EGK_10665; Ig alpha; IGA; Igalpha; ig-alpha; immunoglobulin-associated alpha; LOC100913063; Ly54; Ly-54; MB1; MB-1; MB-1 membrane glycoprotein; Membrane-bound immunoglobulin-associated protein; surface IgM-associated protein | |
Mouse | |
100 Tests | |
Primary | |
Human, Mouse, Rat, Canine, Monkey, Bovine, Pig, Horse, Chicken, Guinea Pig, Baboon, Baboon, Chimpanzee | |
Antibody | |
IgG1 κ |
Safety and Handling
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.