Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD79A Rabbit anti-Bovine, Human, Monkey, Mouse, Pig, Rat, Unconjugated, Clone: IGA/1790R, Abnova™

Catalog No. 89339718 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-339-718 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-339-718 Supplier Abnova Corporation Supplier No. RAB00679
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody.

CD79A Monoclonal antibody specifically detects CD79A in Bovine, Human, Monkey, Mouse, Pig, Rat samples. It is validated for WB-Ce, IHC-P, IF, Flow Cyt.

Specifications

Antigen CD79A
Applications Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence, Flow Cytometry
Classification Monoclonal
Clone IGA/1790R
Conjugate Unconjugated
Description CD79a molecule, immunoglobulin-associated alpha
Gene CD79A
Gene Alias IGA, MB-1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 202-216 of human CD79A
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 973
Target Species Bovine, Human, Monkey, Mouse, Pig, Rat
Content And Storage Store at -20 to -80°C. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.