Learn More
CD79a Recombinant Rabbit Monoclonal Antibody (ZR237), RAbMono™, Zeta
Rabbit Recombinant Monoclonal Antibody Cocktail
Supplier: ZETA CORPORATION Z2533RS

Description
A recommended positive control tissue for this product is Tonsil, however positive controls are not limited to this tissue type. The primary antibody is intended for laboratory professional use in the detection of the corresponding protein in formalin-fixed, paraffin-embedded tissue stained in manual qualitative immunohistochemistry (IHC) testing. This antibody is intended to be used after the primary diagnosis of tumor has been made by conventional histopathology using non-immunological histochemical stains.
CD79a Recombinant Monoclonal antibody specifically detects CD79a in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
CD79a | |
Monoclonal | |
100 μg/mL | |
tris with BSA, NP-40 and <0.1% sodium azide | |
P11912 | |
Rabbit | |
Protein A | |
RUO | |
973 | |
2-8°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
ZR237 | |
Unconjugated | |
CD79A | |
B-cell antigen receptor; B-cell antigen receptor complex-associated protein alpha chain; B-cell antigen receptor complex-associated protein alpha chain-like; CD79; CD79a; CD79A antigen (immunoglobulin-associated alpha); CD79a molecule; CD79a molecule, immunoglobulin-associated alpha; CD79A protein; CD79-alpha protein; EGK_10665; Ig alpha; IGA; Igalpha; ig-alpha; immunoglobulin-associated alpha; LOC100913063; Ly54; Ly-54; MB1; MB-1; MB-1 membrane glycoprotein; Membrane-bound immunoglobulin-associated protein; surface IgM-associated protein | |
ragment (around aa 1-100) of human CD79a protein | |
500 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.