Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD79B Antibody (071), DyLight 550

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290706R
Description
CD79B Monoclonal specifically detects CD79B in Mouse samples. It is validated for ELISA.Specifications
CD79B | |
Monoclonal | |
DyLight 550 | |
B29B-cell-specific glycoprotein B29, B-cell antigen receptor complex-associated protein beta chain, CD79b antigen, CD79B antigen (immunoglobulin-associated beta), CD79b molecule, immunoglobulin-associated beta, IGBAGM6, Ig-beta, Immunoglobulin-associated B29 protein | |
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse CD79B (P15530; Met1-Asp158). | |
0.1 mL | |
Immunology | |
974 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
71 | |
50 mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction