Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD79B Antibody (735451), PE/Cy5.5, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals FAB6814PECY55
Description
CD79B Monoclonal antibody specifically detects CD79B in Mouse samples. It is validated for Flow CytometrySpecifications
CD79B | |
Monoclonal | |
PE-Cyanine5.5 | |
PBS | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Mouse | |
Purified |
Flow Cytometry | |
735451 | |
Flow Cytometry | |
B29B-cell-specific glycoprotein B29, B-cell antigen receptor complex-associated protein beta chain, CD79b antigen, CD79B antigen (immunoglobulin-associated beta), CD79b molecule, immunoglobulin-associated beta, IGBAGM6, Ig-beta, Immunoglobulin-associated B29 protein | |
Chinese hamster ovary cell line CHO-derived recombinant mouse CD79B, Met29-Asp158, Accession # P15530 | |
0.1 mL | |
Immunology | |
974 | |
Store at 4C in the dark. Do not freeze. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction