Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD79B Antibody (HM79-11), mFluor Violet 500 SE, Novus Biologicals™

Armenian Hamster Monoclonal Antibody
Supplier: Novus Biologicals NB10065611MFV500
Description
CD79B Monoclonal antibody specifically detects CD79B in Mouse samples. It is validated for Flow CytometrySpecifications
CD79B | |
Monoclonal | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Armenian Hamster | |
Protein G purified | |
RUO | |
Primary | |
Mouse | |
Purified |
Flow Cytometry | |
HM79-11 | |
Flow Cytometry | |
B29B-cell-specific glycoprotein B29, B-cell antigen receptor complex-associated protein beta chain, CD79b antigen, CD79B antigen (immunoglobulin-associated beta), CD79b molecule, immunoglobulin-associated beta, IGBAGM6, Ig-beta, Immunoglobulin-associated B29 protein | |
CD79 alpha/CD79 beta heterodimer purified from WEH1231 B cells. | |
0.1 mL | |
Immunology | |
974 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction