Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD79B Antibody (IGB/2555), Alexa Fluor™ 647, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308603AF647
Description
CD79B Monoclonal antibody specifically detects CD79B in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin)Specifications
CD79B | |
Monoclonal | |
Alexa Fluor 647 | |
B29B-cell-specific glycoprotein B29, B-cell antigen receptor complex-associated protein beta chain, CD79b antigen, CD79B antigen (immunoglobulin-associated beta), CD79b molecule, immunoglobulin-associated beta, IGBAGM6, Ig-beta, Immunoglobulin-associated B29 protein | |
Recombinant fragment of human CD79B protein (around aa 29-159) (exact sequence is proprietary) (Uniprot: P40259) | |
100 μg | |
Immunology | |
974 | |
Store at 4°C in the dark. | |
IgG1 κ |
ELISA, Immunohistochemistry (Paraffin) | |
IGB/2555 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction