Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD79b Armenian Hamster anti-Mouse, RB780, Clone: HM79b (also known as HM79-16), BD OptiBuild™
Armenian Hamster Monoclonal Antibody
Supplier: BD Biosciences 755347
This item is not returnable.
View return policy
Description
CD79b Monoclonal antibody specifically detects CD79b in Mouse samples. It is validated for Flow Cytometry
Specifications
CD79b | |
Monoclonal | |
0.2 mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Armenian Hamster | |
50 μg | |
Primary | |
Mouse | |
IgG λ1 |
Flow Cytometry | |
HM79b (also known as HM79-16) | |
RB780 | |
Igβ; Igb; Ig-beta; Immunoglobulin-associated beta | |
Mouse B lymphoma, WEHI-123 | |
RUO | |
15985 | |
Store undiluted at 4°C and protected from prolonged exposure to light.Do not freeze.The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography.The antibody was conjugated to the dye under optimum conditions that minimize unconjugated dye and antibody. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction