Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD79b Chimeric Recombinant Rabbit Monoclonal Antibody (HM79-16)
GREENER_CHOICE

Catalog No. PIMA548285
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA548285 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA548285 Supplier Invitrogen™ Supplier No. MA548285
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Specificity: Reacts with an extracellular epitope of CD79b. Preparation: HM79-16 was initially generated by purifying CD79b from WEHI231 B cells and used for immunisation of footpads of an Armenian hamster. Hybridoma supernatants were screened for positive reactivity in surface immunofluorescence of WEHI231 and then the specificity to CD79b was tested by immuno-precipitationRecombinant anti-CD79 antibodies were engineered by cloning and sequencing the full-length IG heavy and light chains of an Armenian hamster (Cricetulus migratorius).

CD79b is the B cell antigen receptor Ig beta chain (BCR-Ig beta), which associates with B cell antigen receptor Ig alpha chain to form a functional heterodimer that interacts with membrane immunoglobulin (mIg). The mIgM component binds antigen and the Ig alpha /I g beta heterodimer tranduces signals.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD79b Chimeric
Applications Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone HM79-16
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% ProClin 300
Gene Cd79b
Gene Accession No. P15530
Gene Alias AGM6; B29; B-cell antigen receptor complex-associated protein beta chain; B-cell-specific glycoprotein B29; CD79; CD79b; CD79B antigen; CD79b molecule; Cd79b molecule, immunoglobulin-associated beta; Ig beta; IGB; Igbeta; Ig-beta; Immunoglobulin-associated B29 protein; immunoglobulin-associated beta; RP23-418O11.7
Gene Symbols Cd79b
Host Species Rabbit
Immunogen HM79-16 was initially generated by purifying CD79b from WEHI231 B cells and used for immunisation of footpads of an Armenian hamster.
Purification Method Protein A
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15985
Target Species Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG λ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.