Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD79B Monoclonal Antibody (OTI7F8), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA810503

Description
CD79b is the B cell antigen receptor Ig beta chain (BCR-Ig beta), which associates with B cell antigen receptor Ig alpha chain to form a functional heterodimer that interacts with membrane immunoglobulin (mIg). The mIgM component binds antigen and the Ig alpha /I g beta heterodimer tranduces signals.Specifications
CD79B | |
Monoclonal | |
Unconjugated | |
CD79B | |
AGM6, B29, IGB | |
Mouse | |
Affinity Chromatography | |
RUO | |
974 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
OTI7F8 | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
P40259 | |
CD79B | |
Full length human recombit protein of human CD79B produced in HEK293T cell. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG2a |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction