Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CD79B Alexa Fluor 594-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB6620T100UG
This item is not returnable.
View return policy
Description
CD79B Monoclonal specifically detects CD79B in Human samples. It is validated for Flow Cytometry.Specifications
CD79B | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
P40259 | |
CD79B | |
Chinese hamster ovary cell line CHO-derived recombinant human CD79B Ala29-Asp159 Accession # P40259 | |
RUO | |
974 | |
Human | |
IgM |
Flow Cytometry | |
683023 | |
Alexa Fluor 594 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
B29, B29B-cell-specific glycoprotein B29, B-cell antigen receptor complex-associated protein beta chain, CD79B, CD79b antigen, CD79B antigen (immunoglobulin-associated beta), CD79b molecule, immunoglobulin-associated beta, IGB, IGBAGM6, Ig-beta, Immunoglobulin-associated B29 protein | |
Mouse | |
100 μg | |
Primary | |
Detects human CD79B in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant mouse CD79B or recombinant human CD79A is observed. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction