Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD79B Mouse anti-Human, Clone: IGB/1842, Novus Biologicals™

Mouse Monoclonal Antibody
$268.00
Specifications
Antigen | CD79B |
---|---|
Clone | IGB/1842 |
Concentration | 0.2 mg/ml |
Dilution | ELISA 2-4 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml |
Applications | ELISA, Immunohistochemistry (Paraffin) |
Description
CD79B Monoclonal specifically detects CD79B in Human samples. It is validated for ELISA, Immunohistochemistry-Paraffin.Specifications
CD79B | |
0.2 mg/ml | |
ELISA, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Human | |
B29B-cell-specific glycoprotein B29, B-cell antigen receptor complex-associated protein beta chain, CD79b antigen, CD79B antigen (immunoglobulin-associated beta), CD79b molecule, immunoglobulin-associated beta, IGBAGM6, Ig-beta, Immunoglobulin-associated B29 protein | |
Recombinant human CD79B protein fragment (around aa29-159) (exact sequence is proprietary) | |
Primary | |
Store at 4°C. |
IGB/1842 | |
ELISA 2-4 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml | |
Monoclonal | |
Purified | |
RUO | |
10 mM PBS with 0.05% BSA | |
974 | |
IgG2b κ | |
Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title