Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD79b Mouse anti-Human, PE, Clone: CB3-1, BD

Catalog No. BDB555679 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB555679 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB555679 Supplier BD Biosciences Supplier No. 555679
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Immunoglobulin (Ig) antigen receptors are composed of a non-covalently- associated complex of Ig and two other proteins, Igα and Igβ, which have been designated in the Fifth International Leukocyte Workshop as CD79a and CD79b respectively. CB3-1 reacts with CD79b, which is expressed on surface Ig(sIg)-positive lymphocytes and B-cell lines but only in the cytoplasm of sIg-negative cells including most terminal deoxynucleotidyl transferase (TdT) positive early pre-B and all cytoplasmic µ positive pre-B cell lines. Antibodies to CD79b are helpful in delineating signal transduction pathways activated via antibody receptors during different stages of B-cell differentiation.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD79b
Applications Flow Cytometry
Classification Monoclonal
Clone CB3-1
Conjugate PE
Description Igβ
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.