Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CD79b Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA582382
Description
Immunogen sequence: DKDDSKAGME EDHTYEGLDI DQTATYEDIV TLRTGEVKWS VGEHPGQE.
CD79b is the B cell antigen receptor Ig beta chain (BCR-Ig beta), which associates with B cell antigen receptor Ig alpha chain to form a functional heterodimer that interacts with membrane immunoglobulin (mIg). The mIgM component binds antigen and the Ig alpha /I g beta heterodimer tranduces signals.
Specifications
CD79b | |
Polyclonal | |
Unconjugated | |
Cd79b | |
AGM6; B29; B-cell antigen receptor complex-associated protein beta chain; B-cell-specific glycoprotein B29; CD79; CD79b; CD79B antigen; CD79b molecule; Cd79b molecule, immunoglobulin-associated beta; Ig beta; IGB; Igbeta; Ig-beta; Immunoglobulin-associated B29 protein; immunoglobulin-associated beta; RP23-418O11.7 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
974 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.10 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P40259 | |
Cd79b | |
Recombinant protein corresponding to Human CD79B. Recombinant protein control fragment (Product #RP-89949). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction