Learn More
CD79B Rabbit anti-Human, Polyclonal , Abnova™
Rabbit polyclonal antibody raised against recombinant human CD79B.
Supplier: Abnova Corporation PAB29311
Description
The B lymphocyte antigen receptor is a multimeric complex that includes the antigen-specific component, surface immunoglobulin (Ig). Surface Ig non-covalently associates with two other proteins, Ig-alpha and Ig-beta, which are necessary for expression and function of the B-cell antigen receptor. This gene encodes the Ig-beta protein of the B-cell antigen component. Alternatively spliced transcript variants encoding different isoforms have been described. [provided by RefSeq]
Sequence: DKDDSKAGMEEDHTYEGLDIDQTATYEDIVTLRTGEVKWSVGEHPGQESpecifications
CD79B | |
Polyclonal | |
Rabbit polyclonal antibody raised against recombinant human CD79B. | |
In PBS, pH 7.2 (40% glycerol, 0.02% sodium azide) | |
CD79B | |
Recombinant protein corresponding to amino acids 182-229 of human CD79B. | |
100 μL | |
Primary | |
Human | |
Liquid |
Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot | |
Unconjugated | |
Immunofluorescence (1-4 ug/mL) Immunohistochemistry (1:200-1:500) Western Blot (1:100-1:250) The optimal working dilution should be determined by the end user. | |
B29/IGB | |
Rabbit | |
Antigen affinity purification | |
RUO | |
974 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.