Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD79B Rabbit anti-Human, Unconjugated, Clone: IGB/2940R, Abnova™

Catalog No. 89339719 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-339-719 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-339-719 Supplier Abnova Corporation Supplier No. RAB00680
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody.

CD79B Monoclonal antibody specifically detects CD79B in Human samples. It is validated for ELISA.

Specifications

Antigen CD79B
Applications ELISA
Classification Monoclonal
Clone IGB/2940R
Conjugate Unconjugated
Description CD79b molecule, immunoglobulin-associated beta
Gene CD79B
Gene Alias B29, IGB
Host Species Rabbit
Immunogen Recombinant protein corresponding to amino acids 29-159 of human CD79B
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 974
Target Species Human
Content And Storage Store at -20 to -80°C. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.