Learn More
CD8 alpha Monoclonal Antibody (53-6.7), Invitrogen™
Rat Monoclonal Antibody
Supplier: Thermo Scientific MA184018
Description
MA1-84018 is recommended for IHC using acetone-fixed frozen tissue sections. For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL. This antibody has been reported to block antigen presentation via MHC class I and inhibit T cell responses to IL-2. This antibody has also been used for depletion of CD8a+ cells. Rat anti Mouse CD8 alpha antibody, clone 53-6.7 detects mouse CD8a, also known as Lyt-2.
CD8 (Cluster of Differentiation 8) is a cell surface glycoprotein found on most cytotoxic T lymphocytes that mediate efficient cell-cell interactions within the immune system. The CD8 antigen acts as a co-receptor with the T-cell receptor on the T lymphocyte to recognize antigens displayed by an antigen presenting cell (APC) in the context of class I MHC molecules. CD8 is found on a T cell subset of normal cytotoxic/suppressor cells which make up approximately 20-35 % of human peripheral blood lymphocytes. The CD8 antigen is also detected on natural killer (NK) cells, subpopulations of peripheral blood null cells, thymocytes and bone marrow cells. The CD8 co-receptor functions as either a homodimer composed of two alpha chains, or as a disulfide-linked heterodimer composed of one alpha and one beta chain. Both alpha and beta chains share significant homology to immunoglobulin variable light chains. The majority of CD8+ T cells express CD8 as a alpha/beta heterodimer. In HIV, the HIV-2 envelope glycoprotein binds CD8 alpha chain (but not the beta chain).
Specifications
CD8 alpha | |
Monoclonal | |
0.5 mg/mL | |
Cd8a | |
CD8, Leu-2, MAL, p32, CD8a | |
Rat | |
Protein G | |
RUO | |
12525 | |
4°C or -20°C if preferred | |
Liquid |
Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation | |
53-6.7 | |
Unconjugated | |
P01731 | |
CD8A | |
Murine thymus or spleen. | |
250 μg | |
Primary | |
Mouse | |
Antibody | |
IgG2a |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.