Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD8 alpha Rat anti-Mouse, Unconjugated, Clone: , MilliporeSigma™

Catalog No. MABF207925 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF2079-25 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABF2079-25 Supplier MilliporeSigma Supplier No. MABF207925UG
Add to Cart
Add to Cart

Rat Monoclonal Antibody

This rat monoclonal Anti-CD8 alpha, clone 53-6.72, Cat. No. MABF2079, is a rat monoclonal antibody that detects CD8 alpha and has been tested for use in Flow Cytometry and functional studies.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD8 alpha
Applications Flow Cytometry, Inhibition Assays
Classification Monoclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified rat monoclonal antibody IgG2a in PBS without azide.
Gene Alias T-cell surface glycoprotein CD8 alpha chain;T-cell surface glycoprotein Lyt-2
Gene Symbols CD8a;Lyt2;Lyt-2
Host Species Rat
Immunogen Murine Thymic/splenic lymphocytes.
Molecular Weight of Antigen 27.5 kDa calculated.
Purification Method Protein G purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Inflammation & Immunology
Primary or Secondary Primary
Test Specificity Clone 53-6.72 is a rat monoclonal antibody that detects CD8 alpha chain in murine thymocytes.
Target Species Mouse
Content And Storage Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Product Type Antibodies
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.