Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD8 Antibody (012), Alexa Fluor™ 350

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290557AF350
Description
CD8 Monoclonal specifically detects CD8 in Mouse samples. It is validated for ELISA.Specifications
CD8 | |
Monoclonal | |
Alexa Fluor 350 | |
CD8, CD8a molecule, LEU2, p32 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse CD8 extracellular domain (NP_001074579.1; Met 1-Tyr 196). | |
0.1 mL | |
Adaptive Immunity, Cytokine Research, Immunology, Innate Immunity, Phospho Specific, Signal Transduction, Stem Cell Markers | |
925 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
12 | |
50 mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction