Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD8 Antibody (12.C7), PerCP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10064021PCP
Description
CD8 Monoclonal antibody specifically detects CD8 in Rabbit samples. It is validated for Flow Cytometry, Product Image.Specifications
CD8 | |
Monoclonal | |
PerCP | |
CD8, CD8a molecule, LEU2, p32 | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Recognizes the rabbit CD8 cell surface antigen, expressed by a subset of T lymphocytes with cytotoxic/suppressor activity. | |
Store at 4C in the dark. | |
IgG1 |
Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
12.C7 | |
Flow Cytometry | |
CD8A | |
Made to Rabbit CD8 | |
0.1 mL | |
Adaptive Immunity, Cytokine Research, Immunology, Innate Immunity, Phospho Specific, Signal Transduction, Stem Cell Markers | |
925 | |
Rabbit | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction