Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD8 Antibody (14), Alexa Fluor™ 532, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250467AF532
Description
CD8 Monoclonal specifically detects CD8 in Human samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry.Specifications
CD8 | |
Monoclonal | |
Alexa Fluor 532 | |
50 mM sodium borate with 0.05% sodium azide | |
CD8A | |
Fibronectin-purified human monocytes | |
0.1 mL | |
925 | |
Human | |
IgG1 |
Flow Cytometry, ELISA, Immunohistochemistry | |
14 | |
Flow Cytometry, ELISA, Immunohistochemistry | |
CD8, CD8a molecule, LEU2, p32 | |
Mouse | |
Protein A purified | |
Primary | |
suitability for western blotting not known | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction