Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD8 Antibody (53-6.7), PE/Cy7, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP149045PECY7
Description
CD8 Monoclonal specifically detects CD8 in Mouse, Rat samples. It is validated for Flow Cytometry.Specifications
CD8 | |
Monoclonal | |
PE-Cyanine7 | |
PBS with 0.05% Sodium Azide | |
CD8A | |
Mouse thymus or spleen. | |
0.1 mL | |
Primary | |
Mouse, Rat | |
IgG2a κ |
Flow Cytometry | |
53-6.7 | |
Flow Cytometry | |
CD8, CD8a molecule, LEU2, p32 | |
Rat | |
Protein A or G purified | |
Adaptive Immunity, Cytokine Research, Immunology, Immunophenotyping, Innate Immunity, Phospho Specific, Signal Transduction, Stem Cell Markers | |
925.0 | |
Store at 4°C in the dark. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction